The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080444.1 | NP_001073913.1 | 8 - 9 | 1237 | 100 |
XM_011529472.1 | XP_011527774.1 | 8 - 9 | 2510 | 100 | |
XM_011529473.1 | XP_011527775.1 | 7 - 8 | 2488 | 100 | |
GenBank mRNA | AK092516.1 | - | 8 - 9 | 1237 | 100 |