The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021078.2 | NP_066564.2 | 7 - 8 | 1238 | 125 |
XM_006721817.3 | XP_006721880.1 | 7 - 8 | 1267 | 125 | |
XM_006721818.3 | XP_006721881.1 | 7 - 8 | 1150 | 125 | |
XM_017024448.1 | XP_016879937.1 | 7 - 8 | 1401 | 125 | |
GenBank mRNA | AF029777.1 | - | 7 - 8 | 1202 | 125 |
BC032743.1 | - | 7 - 8 | 1215 | 125 | |
BC039907.1 | - | 7 - 8 | 1230 | 125 | |
BC105977.1 | - | 7 - 8 | 1207 | 125 |