Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATAD2
Entrez Gene ID
29028
Gene Name
ATPase family, AAA domain containing 2
Gene Aliases
ANCCA, CT137, PRO2000
Chromosome Location
Chr.8: 123319850 - 123396992 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014109.3 NP_054828.2 2 - 3 429 107
XM_011516994.2 XP_011515296.1 2 - 3 956 107
XM_011516995.2 XP_011515297.1 2 - 3 956 107
GenBank mRNA AK097133.1 - 2 - 3 411 107
AL833653.1 - 2 - 3 183 107
AY598335.1 - 2 - 3 321 107
BC113656.1 - 2 - 3 343 107
BP314109.1 - 2 - 3 429 107
CR749832.1 - 3 - 4 686 106

View Full Product Details