Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATAD2
Entrez Gene ID
29028
Gene Name
ATPase family, AAA domain containing 2
Gene Aliases
ANCCA, CT137, PRO2000
Chromosome Location
Chr.8: 123319850 - 123396992 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014109.3 NP_054828.2 8 - 9 1162 78
XM_011516994.2 XP_011515296.1 8 - 9 1689 78
XM_011516995.2 XP_011515297.1 8 - 9 1689 78
GenBank mRNA AK097133.1 - 8 - 9 1144 78
AL833653.1 - 8 - 9 916 78
AY598335.1 - 8 - 9 1054 78
BC113656.1 - 8 - 9 1076 78
CR749832.1 - 9 - 10 1418 78

View Full Product Details