The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014109.3 | NP_054828.2 | 25 - 26 | 3979 | 100 |
XM_011516996.2 | XP_011515298.1 | 14 - 15 | 2399 | 100 | |
GenBank mRNA | AF118088.1 | - | 8 - 9 | 1437 | 100 |
AL833653.1 | - | 25 - 26 | 3733 | 100 | |
AY598335.1 | - | 25 - 26 | 3871 | 100 | |
BC010686.1 | - | 10 - 11 | 1847 | 100 | |
BC019909.1 | - | 7 - 8 | 1183 | 100 | |
BC113656.1 | - | 25 - 26 | 3893 | 100 | |
CR749832.1 | - | 26 - 27 | 4208 | 100 |