The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014109.3 | NP_054828.2 | 27 - 28 | 4237 | 154 |
XM_011516994.2 | XP_011515296.1 | 26 - 27 | 4374 | 154 | |
XM_011516996.2 | XP_011515298.1 | 16 - 17 | 2657 | 154 | |
GenBank mRNA | AF118088.1 | - | 10 - 11 | 1695 | 154 |
AL833653.1 | - | 27 - 28 | 3991 | 154 | |
BC010686.1 | - | 12 - 13 | 2105 | 154 | |
BC019909.1 | - | 9 - 10 | 1441 | 154 | |
BC113656.1 | - | 27 - 28 | 4151 | 154 | |
CR749832.1 | - | 28 - 29 | 4466 | 154 |