The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173982.1 | NP_001167453.1 | 3 - 3 | 5609 | 149 |
NM_018413.5 | NP_060883.1 | 3 - 3 | 5624 | 149 | |
XM_017019369.1 | XP_016874858.1 | 5 - 5 | 5195 | 149 | |
GenBank mRNA | AF131762.1 | - | 1 - 1 | 2353 | 149 |
AL833176.1 | - | 1 - 1 | 3890 | 149 | |
BC030271.1 | - | 1 - 1 | 1873 | 149 |