The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136538.1 | NP_001130010.1 | 10 - 11 | 1536 | 61 |
NM_025247.5 | NP_079523.3 | 9 - 10 | 1443 | 61 | |
GenBank mRNA | AK092356.1 | - | 9 - 10 | 1428 | 61 |
AL832043.1 | - | 10 - 11 | 1514 | 61 | |
AY323912.1 | - | 9 - 10 | 1399 | 61 | |
BC003698.1 | - | 5 - 6 | 600 | 61 | |
BC126358.1 | - | 9 - 10 | 1357 | 61 |