The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270485.1 | NP_001257414.1 | 16 - 17 | 1771 | 72 |
NM_006549.3 | NP_006540.3 | 16 - 17 | 2425 | 72 | |
NM_172216.1 | NP_757365.1 | 15 - 16 | 2296 | 72 | |
XM_005253822.3 | XP_005253879.1 | 15 - 16 | 1598 | 72 | |
XM_011537764.2 | XP_011536066.1 | 16 - 17 | 1773 | 72 | |
XM_017018694.1 | XP_016874183.1 | 17 - 18 | 1781 | 72 | |
XM_017018695.1 | XP_016874184.1 | 17 - 18 | 1990 | 72 | |
XM_017018696.1 | XP_016874185.1 | 17 - 18 | 1832 | 72 | |
XM_017018697.1 | XP_016874186.1 | 17 - 18 | 1854 | 72 | |
XM_017018698.1 | XP_016874187.1 | 16 - 17 | 1778 | 72 | |
XM_017018699.1 | XP_016874188.1 | 16 - 17 | 1652 | 72 | |
XM_017018709.1 | XP_016874198.1 | 15 - 16 | 1693 | 72 | |
GenBank mRNA | AF140507.1 | - | 16 - 17 | 1709 | 72 |
AF287630.1 | - | 16 - 17 | 2419 | 72 | |
AF321386.1 | - | 15 - 16 | 2046 | 72 |