Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNTTIP1
Entrez Gene ID
116092
Gene Name
deoxynucleotidyltransferase, terminal, interacting protein 1
Gene Aliases
C20orf167, Tdif1, dJ447F3.4
Chromosome Location
Chr.20: 45791937 - 45811427 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052951.2 NP_443183.1 12 - 13 918 77
GenBank mRNA AB035676.1 - 12 - 13 913 77
AK314003.1 - 12 - 13 887 77
BC009535.1 - 9 - 10 499 77
BC024290.1 - 12 - 13 863 77
CD678110.1 - 1 - 2 77

View Full Product Details