Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIRPB2
Entrez Gene ID
284759
Gene Name
signal regulatory protein beta 2
Gene Aliases
PTPN1L, PTPNS1L3, dJ776F14.2
Chromosome Location
Chr.20: 1474591 - 1491762 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122962.1 NP_001116434.1 1 - 2 313 91
NM_001134836.1 NP_001128308.1 1 - 2 313 91
XM_005260708.4 XP_005260765.1 1 - 2 122 91
XM_005260709.3 XP_005260766.1 1 - 2 279 91
XM_011529223.2 XP_011527525.1 1 - 2 488 91
GenBank mRNA AK095499.1 - 1 - 2 122 91
AK294440.1 - 1 - 2 139 91
AK297072.1 - 1 - 2 207 91
AY424277.1 - 1 - 2 85 91
BM285394.1 - 1 - 2 315 91
BQ635335.1 - 1 - 2 382 91
DC313096.1 - 1 - 2 211 91
DC358609.1 - 1 - 2 176 91

View Full Product Details