The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042414.2 | NP_001035879.1 | 7 - 8 | 1299 | 73 |
XM_006719844.3 | XP_006719907.1 | 6 - 7 | 1452 | 73 | |
XM_011535137.1 | XP_011533439.1 | 6 - 7 | 1411 | 73 | |
GenBank mRNA | AF448795.1 | - | 6 - 7 | 1162 | 73 |
AK301554.1 | - | 8 - 9 | 1124 | 73 |