The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282447.2 | NP_001269376.1 | 10 - 11 | 1679 | 109 |
NM_025220.4 | NP_079496.1 | 10 - 11 | 1679 | 109 | |
NM_153202.3 | NP_694882.1 | 10 - 11 | 1679 | 109 | |
XM_005260843.1 | XP_005260900.1 | 10 - 11 | 1136 | 109 | |
XM_006723639.1 | XP_006723702.1 | 10 - 11 | 1136 | 109 | |
XM_006723644.2 | XP_006723707.1 | 10 - 11 | 1136 | 109 | |
XM_011529366.1 | XP_011527668.1 | 10 - 11 | 1035 | 109 | |
XM_011529367.1 | XP_011527669.1 | 10 - 11 | 996 | 109 | |
XM_011529368.1 | XP_011527670.1 | 10 - 11 | 1136 | 109 | |
XM_011529370.1 | XP_011527672.1 | 10 - 11 | 1136 | 109 | |
XM_017028083.1 | XP_016883572.1 | 10 - 11 | 1136 | 109 | |
GenBank mRNA | AB055891.1 | - | 10 - 11 | 1080 | 109 |
AF466287.1 | - | 10 - 11 | 998 | 109 | |
AK123015.1 | - | 6 - 7 | 1378 | 109 | |
AK304037.1 | - | 10 - 11 | 1033 | 109 | |
AL117415.1 | - | 1 - 2 | 177 | 109 | |
AY358314.1 | - | 10 - 11 | 1106 | 109 |