Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAM33
Entrez Gene ID
80332
Gene Name
ADAM metallopeptidase domain 33
Gene Aliases
C20orf153, DJ964F7.1
Chromosome Location
Chr.20: 3667973 - 3682131 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282447.2 NP_001269376.1 6 - 7 1287 89
NM_025220.4 NP_079496.1 6 - 7 1287 89
NM_153202.3 NP_694882.1 6 - 7 1287 89
XM_005260843.1 XP_005260900.1 6 - 7 744 89
XM_006723639.1 XP_006723702.1 6 - 7 744 89
XM_006723640.1 XP_006723703.1 6 - 7 744 89
XM_006723644.2 XP_006723707.1 6 - 7 744 89
XM_011529366.1 XP_011527668.1 6 - 7 643 89
XM_011529367.1 XP_011527669.1 6 - 7 604 89
XM_011529368.1 XP_011527670.1 6 - 7 744 89
XM_011529370.1 XP_011527672.1 6 - 7 744 89
XM_011529371.1 XP_011527673.1 6 - 7 744 89
XM_011529373.1 XP_011527675.1 6 - 7 641 89
XM_017028080.1 XP_016883569.1 6 - 7 744 89
XM_017028081.1 XP_016883570.1 6 - 7 665 89
XM_017028082.1 XP_016883571.1 6 - 7 744 89
XM_017028083.1 XP_016883572.1 6 - 7 744 89
GenBank mRNA AF466287.1 - 6 - 7 606 89
AK300429.1 - 6 - 7 655 89
AK304037.1 - 6 - 7 641 89
AY358314.1 - 6 - 7 714 89
BC125113.1 - 6 - 7 648 89

View Full Product Details