The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199513.1 | NP_001186442.1 | 1 - 2 | 49 | 83 |
GenBank mRNA | AL705108.1 | - | 1 - 2 | 49 | 83 |