Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MOCOS
Entrez Gene ID
55034
Gene Name
molybdenum cofactor sulfurase
Gene Aliases
HMCS, MCS, MOS
Chromosome Location
Chr.18: 36187517 - 36268722 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017947.2 NP_060417.2 4 - 5 956 120
GenBank mRNA AK000740.1 - 4 - 5 954 120
AK222886.1 - 4 - 5 943 120
AL834481.1 - 2 - 3 652 120
BC012079.1 - 4 - 5 946 120
BQ931455.1 - 4 - 5 444 120

View Full Product Details