The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133436.3 | NP_597680.2 | 2 - 3 | 475 | 82 |
NM_183356.3 | NP_899199.2 | 3 - 4 | 498 | 82 | |
GenBank mRNA | AK000379.1 | - | 2 - 3 | 443 | 82 |
BC008723.2 | - | 3 - 4 | 325 | 82 | |
DA160359.1 | - | 3 - 4 | 409 | 82 | |
DA961373.1 | - | 2 - 3 | 477 | 82 |