The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014916.3 | NP_055731.2 | 10 - 11 | 1437 | 139 |
XM_011515981.2 | XP_011514283.1 | 10 - 11 | 1448 | 139 | |
GenBank mRNA | AB029002.1 | - | 10 - 11 | 1437 | 139 |
AY130988.1 | - | 10 - 11 | 1144 | 139 |