Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMTK2
Entrez Gene ID
22853
Gene Name
lemur tyrosine kinase 2
Gene Aliases
AATYK2, BREK, KPI-2, KPI2, LMR2, PPP1R100, cprk, hBREK
Chromosome Location
Chr.7: 98106868 - 98209633 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014916.3 NP_055731.2 3 - 4 665 58
XM_011515981.2 XP_011514283.1 3 - 4 676 58
GenBank mRNA AB029002.1 - 3 - 4 665 58
AY130988.1 - 3 - 4 372 58

View Full Product Details