The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199847.1 | NP_001186776.1 | 1 - 2 | 390 | 107 |
NM_020429.2 | NP_065162.1 | 1 - 2 | 390 | 107 | |
NM_181349.2 | NP_851994.1 | 1 - 2 | 390 | 107 | |
XM_017012457.1 | XP_016867946.1 | 1 - 2 | 388 | 107 | |
GenBank mRNA | AB046845.1 | - | 1 - 2 | 358 | 107 |
AF464850.1 | - | 1 - 2 | 50 | 107 | |
BC136804.1 | - | 1 - 2 | 267 | 107 | |
BC144414.1 | - | 1 - 2 | 267 | 107 | |
BC152468.1 | - | 1 - 2 | 358 | 107 | |
DB727777.1 | - | 1 - 2 | 392 | 107 |