The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199847.1 | NP_001186776.1 | 6 - 7 | 818 | 92 |
NM_020429.2 | NP_065162.1 | 6 - 7 | 818 | 92 | |
NM_181349.2 | NP_851994.1 | 6 - 7 | 818 | 92 | |
XM_017012457.1 | XP_016867946.1 | 6 - 7 | 816 | 92 | |
GenBank mRNA | AB046845.1 | - | 6 - 7 | 786 | 92 |
AF199364.2 | - | 6 - 7 | 451 | 92 | |
AF464850.1 | - | 6 - 7 | 478 | 92 | |
BC136804.1 | - | 6 - 7 | 695 | 92 | |
BC144414.1 | - | 6 - 7 | 695 | 92 | |
BC152468.1 | - | 6 - 7 | 786 | 92 |