The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286220.1 | NP_001273149.1 | 6 - 7 | 857 | 67 |
NM_002080.3 | NP_002071.2 | 7 - 8 | 986 | 67 | |
GenBank mRNA | AK098313.1 | - | 7 - 8 | 908 | 67 |
AK223271.1 | - | 7 - 8 | 921 | 67 | |
AK290847.1 | - | 7 - 8 | 919 | 67 | |
AK295993.1 | - | 6 - 7 | 826 | 67 | |
AK302249.1 | - | 6 - 7 | 827 | 67 | |
AK307598.1 | - | 7 - 8 | 873 | 67 | |
BC000525.2 | - | 7 - 8 | 909 | 67 | |
M22632.1 | - | 7 - 8 | 865 | 67 |