The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163315.2 | NP_001156787.2 | 1 - 2 | 1397 | 75 |
XM_005272048.4 | XP_005272105.1 | 1 - 2 | 1391 | 75 | |
XM_005272050.4 | XP_005272107.1 | 1 - 2 | 1391 | 75 | |
XM_011543574.2 | XP_011541876.1 | 1 - 2 | 1391 | 75 | |
XM_011543575.2 | XP_011541877.1 | 1 - 2 | 1391 | 75 | |
XM_011543576.2 | XP_011541878.1 | 1 - 2 | 1391 | 75 | |
XM_011543577.2 | XP_011541879.1 | 1 - 2 | 1391 | 75 | |
XM_011543578.2 | XP_011541880.1 | 1 - 2 | 1391 | 75 | |
XM_011543579.2 | XP_011541881.1 | 1 - 2 | 1391 | 75 | |
XM_011543580.2 | XP_011541882.1 | 1 - 2 | 1391 | 75 | |
XM_017009729.1 | XP_016865218.1 | 1 - 2 | 1391 | 75 | |
GenBank mRNA | AB533280.1 | - | 2 - 3 | 317 | 75 |
AK126722.1 | - | 2 - 3 | 585 | 75 | |
BC126144.1 | - | 1 - 2 | 123 | 75 | |
BC126146.1 | - | 1 - 2 | 123 | 75 |