The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015180.4 | NP_055995.4 | 98 - 99 | 18103 | 117 |
NM_182914.2 | NP_878918.2 | 98 - 99 | 18103 | 117 | |
XM_005267454.1 | XP_005267511.1 | 98 - 99 | 18103 | 117 | |
XM_005267456.1 | XP_005267513.1 | 98 - 99 | 18103 | 117 | |
XM_005267457.1 | XP_005267514.1 | 98 - 99 | 18103 | 117 | |
XM_005267458.1 | XP_005267515.1 | 98 - 99 | 18103 | 117 | |
XM_005267459.1 | XP_005267516.1 | 98 - 99 | 18103 | 117 | |
XM_011536574.1 | XP_011534876.1 | 98 - 99 | 18103 | 117 | |
XM_011536575.2 | XP_011534877.1 | 98 - 99 | 18080 | 117 | |
XM_011536576.2 | XP_011534878.1 | 99 - 100 | 18260 | 117 | |
XM_011536577.2 | XP_011534879.1 | 98 - 99 | 18027 | 117 | |
XM_011536578.1 | XP_011534880.1 | 98 - 99 | 18103 | 117 | |
XM_011536579.1 | XP_011534881.1 | 98 - 99 | 18103 | 117 | |
XM_011536580.1 | XP_011534882.1 | 98 - 99 | 18103 | 117 | |
XM_011536581.1 | XP_011534883.1 | 98 - 99 | 18103 | 117 | |
XM_011536582.1 | XP_011534884.1 | 97 - 98 | 17986 | 117 | |
XM_017021101.1 | XP_016876590.1 | 98 - 99 | 18105 | 117 | |
XM_017021102.1 | XP_016876591.1 | 97 - 98 | 18148 | 117 | |
GenBank mRNA | AB023228.3 | - | 49 - 50 | 7775 | 117 |
AF435011.1 | - | 98 - 99 | 18085 | 117 | |
AF495911.1 | - | 98 - 99 | 18070 | 117 | |
AK090430.1 | - | 16 - 17 | 2479 | 117 | |
AL080133.1 | - | 4 - 5 | 616 | 117 | |
AY061759.1 | - | 46 - 47 | 7406 | 117 | |
BX537642.1 | - | 19 - 20 | 2916 | 117 | |
BX538095.1 | - | 7 - 8 | 1037 | 117 | |
CR749324.1 | - | 18 - 19 | 2800 | 117 |