Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYNE2
Entrez Gene ID
23224
Gene Name
spectrin repeat containing nuclear envelope protein 2
Gene Aliases
EDMD5, NUA, NUANCE, Nesp2, Nesprin-2, SYNE-2, TROPH
Chromosome Location
Chr.14: 63761904 - 64226449 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182910.2 NP_878914.1 7 - 8 1408 73
NM_182914.2 NP_878918.2 114 - 115 20704 73
XM_005267456.1 XP_005267513.1 114 - 115 20746 73
XM_005267459.1 XP_005267516.1 113 - 114 20635 73
XM_011536578.1 XP_011534880.1 114 - 115 20788 73
XM_011536580.1 XP_011534882.1 114 - 115 20746 73
XM_017021104.1 XP_016876593.1 16 - 17 2825 73
GenBank mRNA AB023228.3 - 65 - 66 10376 73
AK001876.1 - 4 - 5 547 73
AK090430.1 - 31 - 32 5011 73
AL117404.1 - 7 - 8 1930 73
AY184204.1 - 7 - 8 1930 73
BX537642.1 - 34 - 35 5449 73
BX640974.1 - 15 - 16 2450 73

View Full Product Details