The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122842.2 | NP_001116314.1 | 7 - 8 | 1073 | 153 |
NM_001199619.1 | NP_001186548.1 | 8 - 9 | 1056 | 153 | |
NM_001199620.1 | NP_001186549.1 | 9 - 10 | 1174 | 153 | |
NM_001199621.1 | NP_001186550.1 | 6 - 7 | 852 | 153 | |
NM_181782.4 | NP_861447.3 | 7 - 8 | 1073 | 153 | |
XM_005266822.4 | XP_005266879.1 | 7 - 8 | 1103 | 153 | |
XM_006715340.3 | XP_006715403.1 | 7 - 8 | 1030 | 153 | |
XM_011535455.2 | XP_011533757.1 | 7 - 8 | 978 | 153 | |
XM_017010269.1 | XP_016865758.1 | 7 - 8 | 789 | 153 | |
XM_017010270.1 | XP_016865759.1 | 8 - 9 | 1035 | 153 | |
XM_017010271.1 | XP_016865760.1 | 7 - 8 | 1033 | 153 | |
XM_017010272.1 | XP_016865761.1 | 7 - 8 | 886 | 153 | |
XM_017010273.1 | XP_016865762.1 | 7 - 8 | 1103 | 153 | |
XM_017010274.1 | XP_016865763.1 | 7 - 8 | 1192 | 153 | |
GenBank mRNA | AB074156.1 | - | 6 - 7 | 601 | 153 |
AB074157.1 | - | 7 - 8 | 873 | 153 | |
AF493978.1 | - | 8 - 9 | 1056 | 153 | |
AK127512.1 | - | 7 - 8 | 883 | 153 | |
AK294558.1 | - | 6 - 7 | 650 | 153 | |
AK307395.1 | - | 8 - 9 | 1035 | 153 | |
AL832621.2 | - | 7 - 8 | 866 | 153 | |
AL832628.2 | - | 7 - 8 | 830 | 153 | |
AL834442.1 | - | 5 - 6 | 494 | 153 | |
BC137094.1 | - | 9 - 10 | 1090 | 153 | |
BC137095.1 | - | 8 - 9 | 972 | 153 | |
BX537385.1 | - | 7 - 8 | 868 | 153 |