The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318746.1 | NP_001305675.1 | 2 - 3 | 314 | 106 |
NM_032730.5 | NP_116119.2 | 2 - 3 | 900 | 106 | |
XM_011536192.2 | XP_011534494.1 | 3 - 4 | 988 | 106 | |
XM_017011375.1 | XP_016866864.1 | 2 - 3 | 900 | 106 | |
XM_017011376.1 | XP_016866865.1 | 2 - 3 | 900 | 106 | |
GenBank mRNA | AF336861.1 | - | 2 - 3 | 474 | 106 |
AK095207.1 | - | 2 - 3 | 314 | 106 | |
AK313962.1 | - | 2 - 3 | 818 | 106 | |
AY063761.1 | - | 2 - 3 | 423 | 106 | |
BC006399.2 | - | 2 - 3 | 695 | 106 |