Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RTN4IP1
Entrez Gene ID
84816
Gene Name
reticulon 4 interacting protein 1
Gene Aliases
NIMP, OPA10
Chromosome Location
Chr.6: 106559237 - 106630921 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF439711.1 - 4 - 5 623 63

View Full Product Details