The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011541706.2 | XP_011540008.1 | 8 - 9 | 1607 | 67 |
GenBank mRNA | AK001632.1 | - | 6 - 7 | 615 | 67 |