The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256865.1 | NP_001243794.1 | 2 - 3 | 350 | 70 |
GenBank mRNA | AK296408.1 | - | 2 - 3 | 304 | 70 |
DC332732.1 | - | 2 - 3 | 350 | 70 |