The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162530.1 | NP_001156002.1 | 10 - 11 | 795 | 105 |
NM_024676.4 | NP_078952.4 | 7 - 8 | 601 | 105 | |
XM_011542153.2 | XP_011540455.1 | 10 - 11 | 840 | 105 | |
XM_017002340.1 | XP_016857829.1 | 10 - 11 | 840 | 105 | |
XM_017002341.1 | XP_016857830.1 | 10 - 11 | 840 | 105 | |
XM_017002342.1 | XP_016857831.1 | 10 - 11 | 840 | 105 | |
XM_017002343.1 | XP_016857832.1 | 10 - 11 | 840 | 105 | |
XM_017002344.1 | XP_016857833.1 | 10 - 11 | 840 | 105 | |
XM_017002345.1 | XP_016857834.1 | 10 - 11 | 840 | 105 | |
GenBank mRNA | AK056459.1 | - | 5 - 6 | 444 | 105 |
AK297015.1 | - | 7 - 8 | 598 | 105 | |
DB034996.1 | - | 6 - 7 | 500 | 105 |