Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SH3D21
Entrez Gene ID
79729
Gene Name
SH3 domain containing 21
Gene Aliases
C1orf113
Chromosome Location
Chr.1: 36306348 - 36329221 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162530.1 NP_001156002.1 1 - 2 28 61
XM_011542153.2 XP_011540455.1 1 - 2 73 61
XM_017002340.1 XP_016857829.1 1 - 2 73 61
XM_017002341.1 XP_016857830.1 1 - 2 73 61
XM_017002342.1 XP_016857831.1 1 - 2 73 61
XM_017002343.1 XP_016857832.1 1 - 2 73 61
XM_017002344.1 XP_016857833.1 1 - 2 73 61
XM_017002345.1 XP_016857834.1 1 - 2 73 61
GenBank mRNA BE537168.1 - 1 - 2 25 61
CN292095.1 - 1 - 2 28 61

View Full Product Details