The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297647.1 | NP_001284576.1 | 14 - 15 | 1386 | 100 |
NM_001297648.1 | NP_001284577.1 | 14 - 15 | 1360 | 100 | |
NM_006314.2 | NP_006305.2 | 14 - 15 | 1365 | 100 | |
XM_011540484.2 | XP_011538786.1 | 14 - 15 | 1345 | 100 | |
XM_011540485.2 | XP_011538787.1 | 11 - 12 | 875 | 100 | |
GenBank mRNA | AB209335.1 | - | 14 - 15 | 1359 | 100 |
AF100153.1 | - | 14 - 15 | 1343 | 100 | |
AK222904.1 | - | 14 - 15 | 1348 | 100 | |
AK292646.1 | - | 14 - 15 | 1347 | 100 | |
BC011604.2 | - | 14 - 15 | 1367 | 100 | |
BC012797.2 | - | 14 - 15 | 1367 | 100 | |
BT006900.1 | - | 14 - 15 | 1328 | 100 |