Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS11
Entrez Gene ID
55823
Gene Name
VPS11, CORVET/HOPS core subunit
Gene Aliases
END1, HLD12, PEP5, RNF108, hVPS11
Chromosome Location
Chr.11: 119067752 - 119081978 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290185.1 NP_001277114.1 3 - 4 760 114
NM_021729.5 NP_068375.3 3 - 4 545 114
XM_011542899.2 XP_011541201.1 3 - 4 503 114
GenBank mRNA AB027508.2 - 3 - 4 515 114
AF308800.1 - 3 - 4 479 114
AK023279.1 - 3 - 4 517 114
AK223158.1 - 3 - 4 505 114
AK302973.1 - 3 - 4 692 114
AL834218.1 - 3 - 4 494 114
BC065563.1 - 3 - 4 479 114
BI561271.1 - 3 - 4 550 114

View Full Product Details