The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170686.1 | NP_001164157.1 | 18 - 19 | 2937 | 83 |
NM_001170687.1 | NP_001164158.1 | 18 - 19 | 2907 | 83 | |
NM_001170688.1 | NP_001164159.1 | 17 - 18 | 2754 | 83 | |
NM_080875.2 | NP_543151.2 | 18 - 19 | 2949 | 83 | |
NR_033183.1 | 18 - 19 | 2732 | 83 | ||
XM_006710372.1 | XP_006710435.1 | 17 - 18 | 2813 | 83 | |
XM_011540731.2 | XP_011539033.1 | 19 - 20 | 3785 | 83 | |
XM_011540736.2 | XP_011539038.1 | 19 - 20 | 3686 | 83 | |
XM_011540737.2 | XP_011539039.1 | 18 - 19 | 3623 | 83 | |
XM_011540741.2 | XP_011539043.1 | 17 - 18 | 3518 | 83 | |
XM_011540742.2 | XP_011539044.1 | 17 - 18 | 3515 | 83 | |
GenBank mRNA | AB074480.1 | - | 18 - 19 | 2860 | 83 |
AB076691.1 | - | 18 - 19 | 2818 | 83 | |
AB076692.1 | - | 17 - 18 | 2665 | 83 | |
AB076693.1 | - | 18 - 19 | 2848 | 83 | |
AB097000.1 | - | 18 - 19 | 2783 | 83 | |
AK091610.1 | - | 12 - 13 | 1753 | 83 | |
AK095914.1 | - | 12 - 13 | 1755 | 83 | |
AK096295.1 | - | 13 - 14 | 1860 | 83 | |
AK097106.1 | - | 14 - 15 | 2179 | 83 | |
AK098785.1 | - | 19 - 20 | 2735 | 83 | |
AK122751.1 | - | 18 - 19 | 2651 | 83 | |
AK122863.1 | - | 17 - 18 | 2943 | 83 | |
AK128553.1 | - | 17 - 18 | 3863 | 83 | |
AK293642.1 | - | 17 - 18 | 2929 | 83 | |
AK302757.1 | - | 18 - 19 | 2901 | 83 | |
AL834527.1 | - | 13 - 14 | 1981 | 83 | |
BC016490.1 | - | 18 - 19 | 2786 | 83 | |
BC037542.1 | - | 18 - 19 | 2829 | 83 |