Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIB2
Entrez Gene ID
142678
Gene Name
mindbomb E3 ubiquitin protein ligase 2
Gene Aliases
ZZANK1, ZZZ5
Chromosome Location
Chr.1: 1615188 - 1630610 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170686.1 NP_001164157.1 2 - 3 327 190
NM_001170687.1 NP_001164158.1 2 - 3 327 148
NM_001170688.1 NP_001164159.1 2 - 3 327 190
NM_001170689.1 NP_001164160.1 2 - 3 344 148
NM_080875.2 NP_543151.2 2 - 3 327 190
XM_006710372.1 XP_006710435.1 2 - 3 299 190
XM_011540731.2 XP_011539033.1 2 - 3 1021 190
XM_011540736.2 XP_011539038.1 2 - 3 1021 190
XM_011540737.2 XP_011539039.1 2 - 3 1021 190
XM_011540741.2 XP_011539043.1 2 - 3 1021 190
XM_011540742.2 XP_011539044.1 2 - 3 1021 190
XM_017000349.1 XP_016855838.1 2 - 3 1021 190
XM_017000350.1 XP_016855839.1 2 - 3 1021 190
XM_017000351.1 XP_016855840.1 2 - 3 1003 190
XM_017000352.1 XP_016855841.1 2 - 3 1021 148
XM_017000353.1 XP_016855842.1 2 - 3 1021 190
XM_017000354.1 XP_016855843.1 2 - 3 1021 190
XM_017000355.1 XP_016855844.1 2 - 3 1021 190
XM_017000356.1 XP_016855845.1 2 - 3 1021 190
XM_017000357.1 XP_016855846.1 2 - 3 1021 190
XM_017000358.1 XP_016855847.1 2 - 3 299 190
XM_017000359.1 XP_016855848.1 2 - 3 299 190
XM_017000360.1 XP_016855849.1 2 - 3 288 148
XM_017000361.1 XP_016855850.1 2 - 3 299 190
XM_017000362.1 XP_016855851.1 2 - 3 339 190
XM_017000363.1 XP_016855852.1 2 - 3 344 148
XM_017000364.1 XP_016855853.1 2 - 3 299 190
GenBank mRNA AB074480.1 - 2 - 3 238 190
AB076691.1 - 2 - 3 238 148
AB076692.1 - 2 - 3 238 190
AB076693.1 - 2 - 3 238 190
AB097000.1 - 2 - 3 203 148
AK098785.1 - 2 - 3 196 190
AK122863.1 - 2 - 3 228 190
AK128167.1 - 2 - 3 304 148
AK128553.1 - 2 - 3 288 148
AK302757.1 - 2 - 3 321 148
AK309362.1 - 1 - 2 305 190
BC037542.1 - 2 - 3 249 148
DA391220.1 - 2 - 3 344 148

View Full Product Details