The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005764.3 | NP_005755.1 | 1 - 2 | 234 | 99 |
GenBank mRNA | BC012303.1 | - | 1 - 2 | 147 | 99 |
BM924665.1 | - | 1 - 2 | 260 | 99 | |
CR450304.1 | - | 1 - 2 | 67 | 99 |