The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144602.3 | NP_653203.1 | 3 - 4 | 521 | 67 |
GenBank mRNA | BC021181.2 | - | 3 - 4 | 432 | 67 |
BC021718.2 | - | 3 - 4 | 432 | 67 |