The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031891.3 | NP_114097.2 | 6 - 7 | 1420 | 111 |
GenBank mRNA | AF217289.1 | - | 5 - 6 | 1034 | 111 |
AK290190.1 | - | 6 - 7 | 1247 | 111 | |
AK313809.1 | - | 6 - 7 | 1299 | 111 | |
BC101047.1 | - | 5 - 6 | 1034 | 111 | |
BC101048.1 | - | 5 - 6 | 1034 | 111 | |
BC101049.1 | - | 5 - 6 | 1034 | 111 |