The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006359.2 | NP_006350.1 | 3 - 4 | 579 | 136 |
XM_011531243.2 | XP_011529545.1 | 4 - 5 | 433 | 136 | |
XM_017029225.1 | XP_016884714.1 | 4 - 5 | 453 | 136 | |
GenBank mRNA | AF030409.1 | - | 3 - 4 | 538 | 136 |
D87743.1 | - | 3 - 4 | 494 | 136 |