The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003080.2 | NP_003071.2 | 6 - 7 | 889 | 83 |
XM_011536079.1 | XP_011534381.1 | 4 - 5 | 362 | 83 | |
GenBank mRNA | AJ222801.1 | - | 6 - 7 | 854 | 83 |
AK313515.1 | - | 6 - 7 | 889 | 83 | |
BC000038.2 | - | 6 - 7 | 761 | 83 |