The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039936.2 | NP_001035025.1 | 2 - 3 | 728 | 116 |
NM_001293069.1 | NP_001279998.1 | 9 - 10 | 999 | 116 | |
NM_001293070.1 | NP_001279999.1 | 9 - 10 | 1213 | 116 | |
NM_001293071.1 | NP_001280000.1 | 7 - 8 | 1118 | 116 | |
NM_001293072.1 | NP_001280001.1 | 8 - 9 | 931 | 116 | |
NM_001293073.1 | NP_001280002.1 | 2 - 3 | 593 | 116 | |
NM_001293075.1 | NP_001280004.1 | 2 - 3 | 593 | 116 | |
NM_001293077.1 | NP_001280006.1 | 2 - 3 | 728 | 116 | |
NM_001293079.1 | NP_001280008.1 | 2 - 3 | 728 | 116 | |
NM_001293080.1 | NP_001280009.1 | 2 - 3 | 728 | 116 | |
NM_001293081.1 | NP_001280010.1 | 2 - 3 | 243 | 116 | |
NM_004067.3 | NP_004058.1 | 8 - 9 | 1174 | 116 | |
XM_011515105.2 | XP_011513407.1 | 11 - 12 | 1054 | 116 | |
XM_011515106.2 | XP_011513408.1 | 10 - 11 | 1015 | 116 | |
XM_011515107.2 | XP_011513409.1 | 10 - 11 | 1797 | 116 | |
XM_017011721.1 | XP_016867210.1 | 12 - 13 | 1072 | 116 | |
XM_017011722.1 | XP_016867211.1 | 11 - 12 | 1815 | 116 | |
GenBank mRNA | AK026415.1 | - | 2 - 3 | 728 | 116 |
AK293959.1 | - | 9 - 10 | 793 | 116 | |
AK294032.1 | - | 8 - 9 | 931 | 116 | |
AK294216.1 | - | 2 - 3 | 243 | 116 | |
AK294271.1 | - | 9 - 10 | 1000 | 116 | |
AK301235.1 | - | 2 - 3 | 252 | 116 | |
AK313021.1 | - | 8 - 9 | 948 | 116 | |
AK316030.1 | - | 2 - 3 | 252 | 116 | |
BC112155.1 | - | 8 - 9 | 919 | 116 | |
EU732752.1 | - | 2 - 3 | 194 | 116 | |
EU732753.1 | - | 2 - 3 | 194 | 116 | |
EU732756.1 | - | 2 - 3 | 329 | 116 | |
EU732757.1 | - | 2 - 3 | 329 | 116 | |
EU732758.1 | - | 2 - 3 | 329 | 116 | |
EU732760.1 | - | 7 - 8 | 681 | 116 | |
EU732762.1 | - | 5 - 6 | 305 | 116 | |
GQ924106.1 | - | 9 - 10 | 1000 | 116 | |
U28926.1 | - | 8 - 9 | 971 | 116 |