Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAM18
Entrez Gene ID
8749
Gene Name
ADAM metallopeptidase domain 18
Gene Aliases
ADAM27, tMDCIII
Chromosome Location
Chr.8: 39584568 - 39730064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320313.1 NP_001307242.1 17 - 18 1999 140
NM_014237.2 NP_055052.1 18 - 19 2071 140
NR_135201.1 16 - 17 1709 140
GenBank mRNA AJ133004.1 - 18 - 19 2016 140
AK313961.1 - 18 - 19 2061 140
AY358321.1 - 17 - 18 1986 140
BC121045.1 - 18 - 19 2016 140
BC121046.1 - 16 - 17 1654 140

View Full Product Details