The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014237.2 | NP_055052.1 | 7 - 8 | 641 | 173 |
GenBank mRNA | AJ133004.1 | - | 7 - 8 | 586 | 173 |
AK313961.1 | - | 7 - 8 | 631 | 173 | |
BC121045.1 | - | 7 - 8 | 586 | 173 |