Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAM18
Entrez Gene ID
8749
Gene Name
ADAM metallopeptidase domain 18
Gene Aliases
ADAM27, tMDCIII
Chromosome Location
Chr.8: 39584568 - 39730064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014237.2 NP_055052.1 8 - 9 707 113
NR_135201.1 7 - 8 529 113
GenBank mRNA AJ133004.1 - 8 - 9 652 113
AK313961.1 - 8 - 9 697 113
BC121045.1 - 8 - 9 652 113
BC121046.1 - 7 - 8 474 113

View Full Product Details