Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAM33
Entrez Gene ID
80332
Gene Name
ADAM metallopeptidase domain 33
Gene Aliases
C20orf153, DJ964F7.1
Chromosome Location
Chr.20: 3667973 - 3682131 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282447.2 NP_001269376.1 17 - 18 2670 81
NM_025220.4 NP_079496.1 17 - 18 2670 81
XM_005260843.1 XP_005260900.1 17 - 18 2127 81
XM_006723639.1 XP_006723702.1 17 - 18 2127 81
XM_006723640.1 XP_006723703.1 17 - 18 2118 81
XM_011529366.1 XP_011527668.1 17 - 18 2026 81
XM_011529367.1 XP_011527669.1 17 - 18 1987 81
XM_011529373.1 XP_011527675.1 16 - 17 2101 81
GenBank mRNA AB055891.1 - 17 - 18 2071 81
AF466287.1 - 17 - 18 1989 81
AK092627.1 - 3 - 4 875 81
AK095953.1 - 9 - 10 869 81
AK123015.1 - 13 - 14 2694 81
AK126487.1 - 11 - 12 3671 81
AY358314.1 - 17 - 18 2097 81
BC125113.1 - 15 - 16 1671 81

View Full Product Details