Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WFDC1
Entrez Gene ID
58189
Gene Name
WAP four-disulfide core domain 1
Gene Aliases
PS20
Chromosome Location
Chr.16: 84294706 - 84329851 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282466.1 NP_001269395.1 3 - 4 690 78
NM_021197.3 NP_067020.2 3 - 4 690 78
GenBank mRNA AF169631.1 - 3 - 4 578 78
AF302109.1 - 3 - 4 601 78
AK075061.1 - 3 - 4 550 78
BC029159.1 - 3 - 4 496 78
CD519223.1 - 3 - 4 604 78

View Full Product Details