Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANGPT4
Entrez Gene ID
51378
Gene Name
angiopoietin 4
Gene Aliases
ANG3, ANG4
Chromosome Location
Chr.20: 869899 - 916367 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322809.1 NP_001309738.1 5 - 6 1074 87
NM_015985.3 NP_057069.1 5 - 6 1074 87
XM_011529239.2 XP_011527541.1 4 - 5 951 87
GenBank mRNA AF074332.1 - 5 - 6 1059 87
AF113708.1 - 5 - 6 955 87
AK304756.1 - 5 - 6 1074 87
BC111976.1 - 5 - 6 1049 87
BC111978.1 - 5 - 6 1049 87

View Full Product Details