The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019030.2 | NP_061903.2 | 22 - 23 | 3558 | 84 |
XM_006714653.2 | XP_006714716.1 | 22 - 23 | 3395 | 84 | |
XM_017009588.1 | XP_016865077.1 | 22 - 23 | 3635 | 84 | |
GenBank mRNA | AF070639.1 | - | 1 - 2 | 85 | 84 |
AK025145.1 | - | 9 - 10 | 1028 | 84 | |
AL079292.1 | - | 12 - 13 | 1760 | 84 | |
AL834496.1 | - | 12 - 13 | 1825 | 84 | |
AY036974.1 | - | 22 - 23 | 3539 | 84 | |
BC056219.1 | - | 22 - 23 | 3535 | 84 | |
BX648101.1 | - | 21 - 22 | 3206 | 84 | |
BX648269.1 | - | 22 - 23 | 3273 | 84 | |
BX648688.1 | - | 15 - 16 | 2460 | 84 | |
BX649135.1 | - | 22 - 23 | 3647 | 84 |