Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D21
Entrez Gene ID
161514
Gene Name
TBC1 domain family member 21
Gene Aliases
MgcRabGAP
Chromosome Location
Chr.15: 73873556 - 73909783 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286434.1 NP_001273363.1 7 - 8 792 106
NM_153356.2 NP_699187.1 8 - 9 900 106
XM_006720409.3 XP_006720472.1 8 - 9 930 106
XM_006720410.3 XP_006720473.1 7 - 8 822 106
XM_006720411.3 XP_006720474.1 6 - 7 718 106
XM_011521281.2 XP_011519583.1 8 - 9 929 106
XM_011521282.2 XP_011519584.1 8 - 9 929 106
XM_011521283.2 XP_011519585.1 8 - 9 929 106
XM_011521284.2 XP_011519586.1 7 - 8 709 106
GenBank mRNA AB449907.1 - 7 - 8 668 106
BC033516.1 - 8 - 9 859 106

View Full Product Details